5-Acetyl-2,4-dimethylthiazole Kosher

Synonyms: 2,4-Dimethyl-5-acetylthiazole, 1-(2,4-Dimethylthiazol-5-yl)ethanone, 1-(2,4-Dimethylthiazol-5-yl)ethan-1-one

W326704-25G-K W326704-100G-K W326704-1KG-K
220233-980EA 201.54 USD
220233-980 220233-978 220233-979
5-Acetyl-2,4-dimethylthiazole Kosher
5-Acetyl-2,4-dimethylthiazole
Formula: C₇H₉NOS
MW: 155.22 g/mol
Boiling Pt: 228 °C (1013 hPa)
Density: 1.15 g/cm³ (20 °C)
MDL Number: MFCD00005331
CAS Number: 38205-60-6
EINECS: 253-826-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR