cis-4-Decenal ≥90% Kosher

Warning

Synonyms: (Z)-4-Decenal

W326402-100G-K W326402-25G-K W326402-1KG-K
220233-969EA 755.38 USD
220233-969 220233-971 220233-970
cis-4-Decenal ≥90% Kosher
cis-4-Decenal
Formula: C₁₀H₁₈O
MW: 154.25 g/mol
Boiling Pt: 78 °C (1013 hPa)
Density: 0.8569 g/cm³ (20 °C)
MDL Number: MFCD00007024
CAS Number: 21662-09-9
EINECS: 244-514-5
UN: 3334
ADR: 9,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR