L(+)-Cysteine ≥97%

Warning

Synonyms: H-Cys-OH, (R)-(+)-Cysteine, (R)-2-Amino-3-mercaptopropanoic acid

W326305-100G W326305-1KG W326305-5KG
220233-965EA 100.92 USD
220233-965 220233-966 220233-967
L(+)-Cysteine ≥97%
L(+)-Cysteine
Formula: HSCH₂CH(NH₂)CO₂H
MW: 121.16 g/mol
Boiling Pt: 254 °C (1013 hPa)
Melting Pt: 115 °C
Density: 1.523 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064306
CAS Number: 52-90-4
EINECS: 200-158-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR