Benzothiazole

Synonyms: Benzo[d]thiazole

W325600-25KG W325600-1KG W325600-10KG
220233-950EA 5938.46 USD
220233-950 220233-949 220233-948
Benzothiazole
Benzothiazole
Formula: C₇H₅NS
MW: 135.19 g/mol
Boiling Pt: 231 °C (1013 hPa)
Density: 1.246 g/cm³ (20 °C)
MDL Number: MFCD00005775
CAS Number: 95-16-9
EINECS: 202-396-2
UN: 2810
ADR: 6.1,III
Merck Index: 13,01108

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR