β-Alanine ≥98%

Synonyms: H-β-Ala-OH, 3-Aminopropionic acid, H-Bala-OH, 3-Aminopropanoic acid

W325201-5KG W325201-10KG W325201-1KG
220233-941EA 80 USD
220233-941 220233-939 220233-940
β-Alanine ≥98%
β-Alanine
Formula: C₃H₇NO₂
MW: 89.09 g/mol
Boiling Pt: 201 °C (1013 hPa)
Melting Pt: 94 °C
Density: 1.44 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008200
CAS Number: 107-95-9
EINECS: 203-536-5
Merck Index: 13,00204

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR