2-Acetylpyridine Kosher

Synonyms: 1-(Pyridin-2-yl)ethanone, Methyl 2-pyridyl ketone

W325104-25KG-K W325104-1KG-K W325104-5KG-K W325104-100G-K
220233-936EA 5015.38 USD
220233-936 220233-935 220233-937 220233-933
2-Acetylpyridine Kosher
2-Acetylpyridine
Formula: C₇H₇NO
MW: 121.14 g/mol
Boiling Pt: 192.3 °C (1013 hPa)
Melting Pt: ∼10 °C
Density: 1.0818 g/cm³ (20 °C)
Flash Pt: 75 °C
Storage Temperature: Ambient
MDL Number: MFCD00006303
CAS Number: 1122-62-9
EINECS: 214-355-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR