2,6-Xylenol ≥99%

Danger

Synonyms: 2,6-Dimethylphenol, 2-Hydroxy-m-xylene, 2-Hydroxy-3-xylene

W324906-1KG W324906-10KG W324906-5KG
220233-926EA 292.31 USD
220233-926 220233-925 220233-927
2,6-Xylenol ≥99%
2,6-Xylenol
Formula: C₈H₁₀O
MW: 122.17 g/mol
Boiling Pt: 203 °C (1013 hPa)
Melting Pt: 41 - 42 °C
Density: 1.13 g/cm³ (20 °C)
Flash Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD00002240
CAS Number: 576-26-1
EINECS: 209-400-1
UN: 2261
ADR: 6.1,II
Merck Index: 12,10215

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR