2,3,5-Trimethylpyrazine Kosher

W324426-1KG-K W324426-100G-K
220233-914EA 6969.23 USD
220233-914 220233-913
2,3,5-Trimethylpyrazine Kosher
2,3,5-Trimethylpyrazine
Formula: C₇H₁₀N₂
MW: 122.17 g/mol
Boiling Pt: 171 °C (1013 hPa)
Density: 0.9958 g/cm³ (0 °C)
MDL Number: MFCD00006145
CAS Number: 14667-55-1
EINECS: 238-712-0
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR