trans-2-Nonenal ≥93% Kosher

Warning

Synonyms: (E)-2-Nonenal

W321303-1KG-K W321303-200G-K W321303-4KG-K
220233-527EA 881.54 USD
220233-527 220233-528 220233-529
trans-2-Nonenal ≥93% Kosher
trans-2-Nonenal
Formula: C₉H₁₆O
MW: 140.23 g/mol
Boiling Pt: 100 °C (1013 hPa)
Density: 0.8505 g/cm³ (15 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00007012
CAS Number: 18829-56-6
EINECS: 242-609-6
Merck Index: 12,06771

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR