2-Acetyl-1H-pyrrole

Synonyms: Methyl-2-pyrrolylketon, Methyl 2-pyrrolyl ketone, 1-(1H-Pyrrol-2-yl)ethanone, 1-(1H-Pyrrol-2-yl)ethan-1-one, 2-Acetylpyrrole

W320218-1KG-K W320218-100G-K W320218-5KG-K
220232-011EA 1569.23 USD
220232-011 220232-010 220232-012
2-Acetyl-1H-pyrrole
2-Acetyl-1H-pyrrole
Formula: C₆H₇NO
MW: 109.13 g/mol
Boiling Pt: 220 °C (1013 hPa)
Melting Pt: 63 - 65 °C
MDL Number: MFCD00005220
CAS Number: 1072-83-9
EINECS: 214-016-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR