1-Methylnaphthalene Kosher

Warning

Synonyms: α-Methylnaphthalene

W319309-5KG-K W319309-100G-K W319309-1KG-K
220233-825EA 723.08 USD
220233-825 220233-823 220233-824
1-Methylnaphthalene Kosher
1-Methylnaphthalene
Formula: C₁₁H₁₀
MW: 142.2 g/mol
Boiling Pt: 244…245 °C (1013 hPa)
Melting Pt: –33…–32 °C
Density: 1.022 g/cm³ (20 °C)
Flash Pt: 122 °C
Storage Temperature: Ambient
MDL Number: MFCD00004034
CAS Number: 90-12-0
EINECS: 201-966-8
UN: 3082
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR