4-Methylbiphenyl ≥98% Kosher

Synonyms: 4-Methyl-1,1'-biphenyl, 4-Phenyltoluene

W318604-100G-K W318604-25G-K W318604-1KG-K
220233-809EA 2630.77 USD
220233-809 220233-811 220233-810
4-Methylbiphenyl ≥98% Kosher
4-Methylbiphenyl
Formula: C₁₃H₁₂
MW: 168.24 g/mol
Boiling Pt: 267 °C (1013 hPa)
Melting Pt: 47…48 °C
Density: 1.015 g/cm³ (27 °C)
MDL Number: MFCD00008544
CAS Number: 644-08-6
EINECS: 211-409-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR