2-Methoxy-3-methylpyrazine, halal

W318302-5KG-K W318302-1KG-K W318302-250G-K
220233-803EA 7276.92 USD
220233-803 220233-800 220233-801
2-Methoxy-3-methylpyrazine, halal
2-Methoxy-3-methylpyrazine
Formula: C₆H₈N₂O
MW: 124.14 g/mol
Boiling Pt: 87 °C (60 mmHg)
Density: 1.07 g/cm³ (20 °C)
MDL Number: MFCD00006127
CAS Number: 2847-30-5
EINECS: 220-651-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR