trans-3-Hexenoic acid ≥98% Kosher

W317004-5KG-K W317004-100G-K W317004-1KG-K
220233-755EA 1086.15 USD
220233-755 220233-753 220233-754
trans-3-Hexenoic acid ≥98% Kosher
trans-3-Hexenoic acid
Formula: C₆H₁₀O₂
MW: 114.14 g/mol
Boiling Pt: 209 °C (1013 hPa)
Melting Pt: 14 °C
Density: 0.965 g/cm³ (20 °C)
MDL Number: MFCD00002786
CAS Number: 1577-18-0
EINECS: 216-417-8
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR