trans-2-Hexenoic acid ≥98%, FG

Synonyms: trans-Hex-2-enoic acid

W316903-100G-K W316903-1KG-K W316903-5KG-K
220233-749EA 220 USD
220233-749 220233-750 220233-751
trans-2-Hexenoic acid ≥98%, FG
trans-2-Hexenoic acid
Formula: C₆H₁₀O₂
MW: 114.14 g/mol
Boiling Pt: 217 °C (1013 hPa)
Melting Pt: 32 °C
Density: 0.946 g/cm³ (45 °C)
MDL Number: MFCD00002705
CAS Number: 13419-69-7
EINECS: 236-528-5
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR