4-Ethylphenol ≥98% Kosher

Warning

W315605-10KG-K W315605-1KG-K
220233-708EA 2984.62 USD
220233-708 220233-709
4-Ethylphenol ≥98% Kosher
4-Ethylphenol
Formula: C₈H₁₀O
MW: 122.17 g/mol
Boiling Pt: 218 °C (1013 hPa)
Melting Pt: 40…43 °C
Density: 1.011 g/cm³ (20 °C)
Flash Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00002393
CAS Number: 123-07-9
EINECS: 204-598-6
UN: 2430
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR