(±)-2-Ethyl-1-hexanol ≥99%

Warning

Synonyms: 2-Ethylhexan-1-ol

W315109-8KG-K W315109-1KG-K W315109-4KG-K
220233-686EA 470.77 USD
220233-686 220232-873 220232-874
(±)-2-Ethyl-1-hexanol ≥99%
(±)-2-Ethyl-1-hexanol
Formula: CH₃(CH₂)₃CH(C₂H₅)CH₂OH
MW: 130.23 g/mol
Boiling Pt: 183 °C (1013 hPa)
Melting Pt: –76 °C
Density: 0.83 g/cm³ (20 °C)
Flash Pt: 81 °C
Storage Temperature: Ambient
MDL Number: MFCD00004746
CAS Number: 104-76-7
EINECS: 203-234-3
REACH: 01-2119487289-20
Merck Index: 13,03842
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cc81f9275fbd3a7f8fb0e453419eab1652a4fdec&vid=1f7059eb6b27d30ac2c7e6b83f6bc0dff6002fe7&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR