2,6-Dimethoxyphenol ≥97% Kosher

Warning

W313718-100G-K W313718-5KG-K W313718-1KG-K
220233-677EA 212.31 USD
220233-677 220233-681 220233-679
2,6-Dimethoxyphenol ≥97% Kosher
2,6-Dimethoxyphenol
Formula: C₈H₁₀O₃
MW: 154.17 g/mol
Boiling Pt: 261 °C (1013 hPa)
Melting Pt: 52 - 56 °C
Storage Temperature: Ambient
MDL Number: MFCD00064434
CAS Number: 91-10-1
EINECS: 202-041-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR