Butylamine ≥99% Kosher

Danger

Synonyms: 1-Butylamine, 1-Aminobutane, n-Butylamine

W313009-4KG-K W313009-20KG-K W313009-1KG-K
220233-653EA 263.08 USD
220233-653 220233-652 220233-651
Butylamine ≥99% Kosher
Butylamine
Formula: H₃C(CH₂)₂CH₂NH₂
MW: 73.14 g/mol
Boiling Pt: 77 °C (1013 hPa)
Melting Pt: –50 °C
Density: 0.741 g/cm³ (20 °C)
Flash Pt: –8 °C
Storage Temperature: Ambient
MDL Number: MFCD00011690
CAS Number: 109-73-9
EINECS: 203-699-2
UN: 1125
ADR: 3,II
Merck Index: 12,01578

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR