Veratraldehyde ≥98% Kosher

Warning

Synonyms: 3,4-Dimethoxybenzaldehyde

W310905-10KG-K W310905-25KG-K W310905-5KG-K W310905-1KG-K
220233-513EA 1192.31 USD
220233-513 220231-155 220231-156 220233-514
Veratraldehyde ≥98% Kosher
Veratraldehyde
Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Boiling Pt: 281 °C (1013 hPa)
Melting Pt: 40…43 °C
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00003363
CAS Number: 120-14-9
EINECS: 204-373-2
Merck Index: 12,10084

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR