4-Acetoxy-3-methoxybenzaldehyde ≥98% Kosher

W310808-100G-K W310808-5KG-K W310808-1KG-K
220233-509EA 107.69 USD
220233-509 220233-511 220233-510
4-Acetoxy-3-methoxybenzaldehyde ≥98% Kosher
4-Acetoxy-3-methoxybenzaldehyde
Formula: C₁₀H₁₀O₄
MDL Number: MFCD00003362
CAS Number: 881-68-5
EINECS: 212-920-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR