Vanillin ≥97%, FG

Warning

Synonyms: 4-Hydroxy-3-methoxybenzaldehyde

W310700-1KG-K W310700-25KG-K W310700-10KG-K
220233-009EA 312.31 USD
220233-009 220233-010 220233-008
Vanillin ≥97%, FG
Vanillin
Formula: C₈H₈O₃
MW: 152.15 g/mol
Boiling Pt: 285 °C (1013 hPa)
Melting Pt: ∼82 °C
Density: 1.056 g/cm³ (20 °C)
Flash Pt: 153 °C
Storage Temperature: Ambient
MDL Number: MFCD00006942
CAS Number: 121-33-5
EINECS: 204-465-2
Merck Index: 13,09998
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd6583c5aea50cd090e103650a05eab491b4d38e&vid=535f9a4c80d51d7343126ae5e79c6c83b3beefa7&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR