(±)-γ-Valerolactone, halal

Synonyms: (±)-4-Hydroxypentanoic acid lactone, (±)-Pentan-4-olide, γ-Pentanolactone, 5-Methyldihydrofuran-2(3H)-one, gamma-Valerolactone

W310301-10KG-K W310301-5KG-K W310301-1KG-K
220233-004EA 1526.15 USD
220233-004 220233-006 220233-005
(±)-γ-Valerolactone, halal
(±)-γ-Valerolactone
Formula: C₅H₈O₂
MW: 100.12 g/mol
Boiling Pt: 207 °C (1013 hPa)
Melting Pt: –31 °C
Density: 1.05 g/cm³ (20 °C)
MDL Number: MFCD00005400
CAS Number: 108-29-2
EINECS: 203-569-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR