Isovaleric acid ≥99% FCC

Danger

Synonyms: 3-Methylbutyric acid, Isopropyl acetic acid, 3-Methylbutanoic acid

W310204-10KG W310204-1KG W310204-20KG W310204-5KG
220232-994EA 516.92 USD
220232-994 220232-995 220232-996 220232-997
Isovaleric acid ≥99% FCC
Isovaleric acid
Formula: C₅H₁₀O₂
MW: 102.13 g/mol
Boiling Pt: 177 °C (1013 hPa)
Melting Pt: –33 °C
Density: 0.93 g/cm³ (20 °C)
Flash Pt: 74 °C
Storage Temperature: Ambient
MDL Number: MFCD00002726
CAS Number: 503-74-2
EINECS: 207-975-3
UN: 3265
ADR: 8,II
Merck Index: 12,05251

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR