1-Undecanol ≥97% Kosher

Synonyms: 1-Undecyl alcohol, Undecyl alcohol, Undecan-1-ol, Decyl carbinol, 1-Hydroxy undecane

W309702-4KG-K W309702-8KG-K W309702-1KG-K
220233-494EA 740 USD
220233-494 220233-495 220233-493
1-Undecanol ≥97% Kosher
1-Undecanol
Formula: C₁₁H₂₄O
MW: 172.31 g/mol
Boiling Pt: 243 °C (1013 hPa)
Melting Pt: 13…15 °C
Density: 0.83 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00004751
CAS Number: 112-42-5
EINECS: 203-970-5
UN: 3082
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR