10-Undecenal ≥90% Kosher

Synonyms: Undecylenic aldehyde, Undec-10-enal, 10-Undecenyl aldehyde, 10-Undecenal

W309508-100G-K W309508-1KG-K W309508-4KG-K
220233-489EA 100.92 USD
220233-489 220233-490 220233-491
10-Undecenal ≥90% Kosher
10-Undecenal
Formula: C₁₁H₂₀O
MW: 168.28 g/mol
Boiling Pt: 240 °C (1013 hPa)
Melting Pt: 7 °C
Density: 0.8467 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00007032
CAS Number: 112-45-8
EINECS: 203-973-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR