2-Undecanone ≥97% FCC

Synonyms: Methyl nonyl ketone, Nonyl methyl ketone

W309311-1KG-K W309311-100G-K
220233-486EA 1538.46 USD
220233-486 220233-485
2-Undecanone ≥97% FCC
2-Undecanone
Formula: C₁₁H₂₂O
MW: 170.3 g/mol
Boiling Pt: 231 °C (1013 hPa)
Melting Pt: ~ 12 °C
Density: 0.83 g/cm³ (20 °C)
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00009583
CAS Number: 112-12-9
EINECS: 203-937-5
UN: 3082
ADR: 9,III
Merck Index: 12,06178

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR