(±)-γ-Undecalactone Kosher

Synonyms: (±)-Undecan-4-olide, Gamma-undecalactone, Undecanoic gamma-Lactone

W309109-10KG-K W309109-1KG-K W309109-5KG-K
220232-542EA 816.92 USD
220232-542 220232-543 220232-544
(±)-γ-Undecalactone Kosher
(±)-γ-Undecalactone
Formula: C₁₁H₂₀O₂
MW: 184.28 g/mol
Boiling Pt: 297 °C (1013 hPa)
Density: 0.9414 g/cm³ (20 °C)
MDL Number: MFCD00005405
CAS Number: 104-67-6
EINECS: 203-225-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR