Thymol ≥99%

Danger

Synonyms: 4-Isopropyl-3-hydroxytoluene, 2-Isopropyl-5-methylphenol, 6-Isopropyl-m-cresol

W306606-10KG-K W306606-1KG-K
220233-445EA 647.69 USD
220233-445 220233-446
Thymol ≥99%
Thymol
Formula: 2-[(CH₃)₂CH]C₆H₃-5-(CH₃)OH
MW: 150.22 g/mol
Boiling Pt: 233 °C (1013 hPa)
Melting Pt: 49…51 °C
Density: 0.965 g/cm³ (25 °C)
Flash Pt: 104 °C
Storage Temperature: Ambient
MDL Number: MFCD00002309
CAS Number: 89-83-8
EINECS: 201-944-8
UN: 2430
ADR: 8,III
Merck Index: 13,09476
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3b3268b34e160525798cab810087412f9b9a9db6&vid=7293cafb537e97291a403e856ca7fdb0223ba714&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR