Sodium benzoate ≥99%

Synonyms: Benzoic acid sodium salt

W302503-10KG-K W302503-1KG-K
220233-333EA 76.92 USD
220233-333 220233-334
Sodium benzoate ≥99%
Sodium benzoate
Formula: C₆H₅COONa
MW: 144.11 g/mol
Melting Pt: 410…430 °C
Density: 1.44 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00012463
CAS Number: 532-32-1
EINECS: 208-534-8
REACH: 01-2119460683-35
Merck Index: 13,08654

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR