Salicylaldehyde ≥98%

Warning

Synonyms: 2-Hydroxybenzaldehyde

W300403-10KG-K W300403-1KG-K W300403-25KG-K
220233-323EA 609.23 USD
220233-323 220233-324 220231-998
Salicylaldehyde ≥98%
Salicylaldehyde
Formula: C₇H₆O₂
MW: 122.12 g/mol
Boiling Pt: 196 °C (1013 hPa)
Melting Pt: -7 °C
Density: 1.17 g/cm³ (20 °C)
Flash Pt: 79 °C
Storage Temperature: Ambient
MDL Number: MFCD00003317
CAS Number: 90-02-8
EINECS: 201-961-0
Merck Index: 13,08405

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR