(-)-Quinine hydrochloride dihydrate Kosher

Danger

Synonyms: Quinine hydrochloride dihydrate, Quinine monohydrochloride dihydrate, (R)-(6-Methoxyquinolin-4-yl)((1S,2S,4S,5R)-5-vinylquinuclidin-2-yl)methanol hydrochloride dihydrate

W297607-5KG-K W297607-100G-K W297607-1KG-K
220233-294EA 3415.38 USD
220233-294 220233-292 220233-293
(-)-Quinine hydrochloride dihydrate Kosher
(-)-Quinine hydrochloride dihydrate
Formula: C₂₀H₂₅ClN₂O₂·2H₂O
MW: 396.91 g/mol
Melting Pt: 158 - 160 °C
Flash Pt: 122 °C
Storage Temperature: Ambient
MDL Number: MFCD00151248
CAS Number: 6119-47-7
EINECS: 205-001-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR