Propyl-3,4,5-trihydroxybenzoate ≥98% FCC

Warning

Synonyms: Gallic acid propyl ester, Propyl gallate, Propyl 3,4,5-trihydroxybenzoate, Progalline P, N-Propyl gallate

W294705-10KG-K W294705-1KG-K
220229-758EA 1530.77 USD
220229-758 220229-759
Propyl-3,4,5-trihydroxybenzoate ≥98% FCC
Propyl-3,4,5-trihydroxybenzoate
Formula: C₁₀H₁₂O₅
MW: 212.2 g/mol
Boiling Pt: 449 °C (1013 hPa)
Melting Pt: 145…148 °C
Density: 1.21 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002196
CAS Number: 121-79-9
EINECS: 204-498-2
Merck Index: 12,08044

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR