4-Isopropylbenzyl alcohol ≥97% Kosher

Warning

Synonyms: (4-Isopropylphenyl)methanol

W293318-100G-K W293318-1KG-K
220233-247EA 144.31 USD
220233-247 220233-248
4-Isopropylbenzyl alcohol ≥97% Kosher
4-Isopropylbenzyl alcohol
Formula: C₁₀H₁₄O
MW: 150.22 g/mol
Boiling Pt: 249 °C (1013 hPa)
Melting Pt: 28 °C
Density: 0.978 g/cm³ (20 °C)
MDL Number: MFCD00004663
CAS Number: 536-60-7
EINECS: 208-640-4
Merck Index: 13,02646

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR