Propionic acid ≥99%, FG

Danger

Synonyms: Propanoic acid

W292419-100G-K W292419-5KG-K W292419-1KG-K
220232-845EA 120.62 USD
220232-845 220233-215 220232-846
Propionic acid ≥99%, FG
Propionic acid
Formula: C₃H₆O₂
MW: 74.08 g/mol
Boiling Pt: 141 °C (1013 hPa)
Melting Pt: –21 °C
Density: 0.99 g/cm³ (20 °C)
Flash Pt: 49 °C
Storage Temperature: Ambient
MDL Number: MFCD00002756
CAS Number: 79-09-4
EINECS: 201-176-3
UN: 3463
ADR: 8,II
REACH: 01-2119486971-24
Merck Index: 13,07917

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR