Propionaldehyde ≥97% Kosher

Danger

Synonyms: Propanal

W292303-1KG-K W292303-20KG-K W292303-4KG-K
220232-836EA 167.69 USD
220232-836 220232-837 220232-838
Propionaldehyde ≥97% Kosher
Propionaldehyde
Formula: C₃H₆O
MW: 58.08 g/mol
Boiling Pt: 48 °C (1013 hPa)
Melting Pt: –81 °C
Density: 0.811 g/cm³ (20 °C)
Flash Pt: –40 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007020
CAS Number: 123-38-6
EINECS: 204-623-0
UN: 1275
ADR: 3,II
Merck Index: 13,07915

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR