Polysorbate 80 Kosher

Synonyms: Sorbitan monooleate, ethoxylated, Polyoxyethylene-80-Sorbitan Monolaurate, Polyoxyethylene (20) sorbitan monooleate

W291706-25KG-K W291706-10KG-K W291706-1KG-K
220231-425EA 721.54 USD
220231-425 220231-423 220231-424
Polysorbate 80 Kosher
Polysorbate 80
Melting Pt: 100 °C
Flash Pt: 113 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00082107
CAS Number: 9005-65-6
EINECS: 500-019-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR