Polysorbate 60

Synonyms: Sorbitan monostearate, ethoxylated, Polyoxyethylene sorbitan monostearate

W291609-1KG W291609-10KG
220233-212EA 80 USD
220233-212 220233-211
Polysorbate 60
Polysorbate 60
Formula: C₆₄H₁₂₆O₂₆
Storage Temperature: Ambient
MDL Number: MFCD01780110
CAS Number: 9005-67-8
EINECS: 500-020-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR