Polysorbate 20

W291501-1KG W291501-25KG W291501-10KG
220233-208EA 117.69 USD
220233-208 220233-209 220233-207
Polysorbate 20
Polysorbate 20
Formula: C₅₈H₁₁₄O₂₆
Storage Temperature: Ambient
MDL Number: MFCD00165986
CAS Number: 9005-64-5
EINECS: 500-018-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR