1,1-Dimethoxy-2-phenylethane ≥98%

Synonyms: Phenylacetaldehyde dimethyl acetal

W287601-1KG-K W287601-5KG-K W287601-10KG-K
220233-119EA 80 USD
220233-119 220233-120 220233-118
1,1-Dimethoxy-2-phenylethane ≥98%
1,1-Dimethoxy-2-phenylethane
Formula: C₆H₅CH₂CH(OCH₃)₂
MW: 166.22 g/mol
Boiling Pt: 221 °C (1013 hPa)
Melting Pt: 62…64 °C
Density: 1 g/cm³ (20 °C)
MDL Number: MFCD00008487
CAS Number: 101-48-4
EINECS: 202-945-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR