Phenethyl benzoate ≥99%

W286001-10KG-K W286001-5KG-K W286001-1KG-K
220232-986EA 80 USD
220232-986 220232-988 220232-987
Phenethyl benzoate ≥99%
Phenethyl benzoate
Formula: C₁₅H₁₄O₂
MW: 226.27 g/mol
CAS Number: 94-47-3
EINECS: 202-336-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR