2-Phenylethanol ≥99% FCC

Danger

Synonyms: Phenethyl alcohol, Benzylcarbinol, β-Hydroxyethylbenzene, Benzyl carbinol, Benzenethanol

W285811-100G-K W285811-1KG-K W285811-25G-K
220231-663EA 396.92 USD
220231-663 220231-664 220231-665
2-Phenylethanol ≥99% FCC
2-Phenylethanol
Formula: C₆H₅CH₂CH₂OH
MW: 122.17 g/mol
Boiling Pt: 432 °C (1013 hPa)
Melting Pt: 103…105 °C
Density: 1.02 g/cm³ (20 °C)
Flash Pt: 102 °C
Storage Temperature: Ambient
MDL Number: MFCD00002886
CAS Number: 60-12-8
EINECS: 200-456-2
UN: 2810
ADR: 6.1,III
Merck Index: 12,07370

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR