4-Pentenoic acid ≥98% Kosher

Synonyms: Allylacetic acid, Pent-4-enoic acid

W284300-25G-K W284300-1KG-K W284300-100G-K
220232-957EA 139.23 USD
220232-957 220232-956 220232-955
4-Pentenoic acid ≥98% Kosher
4-Pentenoic acid
Formula: C₅H₈O₂
MW: 100.12 g/mol
Boiling Pt: 187 °C (1013 hPa)
Melting Pt: –18 °C
Density: 0.98 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004408
CAS Number: 591-80-0
EINECS: 209-732-7
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR