2-Pentanone ≥98%

Danger

Synonyms: Methyl Propyl Ketone

W284203-1KG W284203-20KG W284203-4KG W284203-100G
220232-950EA 166.15 USD
220232-950 220232-951 220232-952 220232-949
2-Pentanone ≥98%
2-Pentanone
Formula: C₅H₁₀O
MW: 86.13 g/mol
Boiling Pt: 100 °C (1013 hPa)
Melting Pt: –78 °C
Density: 0.809 g/cm³ (20 °C)
Flash Pt: 7 °C
Storage Temperature: Ambient
MDL Number: MFCD00009400
CAS Number: 107-87-9
EINECS: 203-528-1
UN: 1249
ADR: 3,II
Merck Index: 12,06193

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR