Pentadecan-15-olide Kosher

Synonyms: Cyclopentadecanolide, Oxacyclohexadecan-2-one, Pentadecanolide, ω-Pentadecalactone, omega-Pentadecalactone, Oxa-2-cyclohexadecanone

W284009-100G-K W284009-5KG-K W284009-1KG-K
220231-413EA 147.38 USD
220231-413 220231-415 220231-414
Pentadecan-15-olide Kosher
Pentadecan-15-olide
Formula: C₁₅H₂₈O₂
MW: 240.39 g/mol
MDL Number: MFCD00039667
CAS Number: 106-02-5
EINECS: 203-354-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR