Palmitic acid, FG

Synonyms: Hexadecanoic acid, Hexadecylic acid

W283207-25KG-K W283207-10KG-K W283207-1KG-K
220232-833EA 440 USD
220232-833 220232-831 220232-832
Palmitic acid, FG
Palmitic acid
Formula: C₁₆H₃₂O₂
MW: 256.43 g/mol
Boiling Pt: 352 °C (1013 hPa)
Melting Pt: 61 - 63 °C
Density: 0.853 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002747
CAS Number: 57-10-3
EINECS: 200-312-9
Merck Index: 13,07064

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR