(±)-2-Octanol ≥97%, halal

Danger

Synonyms: (±)-2-Hydroxy octane, 2-Capryl alcohol, (±)-Methyl hexyl carbinol, (±)-Hexyl methyl carbinol, DL-2-Octanol, Octan-2-ol

W280100-1KG-K W280100-8KG-K
220232-484EA 117.69 USD
220232-484 220232-486
(±)-2-Octanol ≥97%, halal
(±)-2-Octanol
Formula: C₈H₁₈O
MW: 130.23 g/mol
Boiling Pt: 180 °C (1013 hPa)
Melting Pt: –39 °C
Density: 0.818 g/cm³ (20 °C)
Flash Pt: 55 °C
Storage Temperature: Ambient
MDL Number: MFCD00004591
CAS Number: 123-96-6
EINECS: 204-667-0
UN: 1987
ADR: 3,III
Merck Index: 13,06785

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR