1-Octanol ≥98% Kosher

Warning

Synonyms: n-Octanol

W280011-4KG-K W280011-8KG-K W280011-1KG-K
220232-749EA 618.46 USD
220232-749 220232-750 220232-748
1-Octanol ≥98% Kosher
1-Octanol
Formula: H₃C(CH₂)₆CH₂OH
MW: 130.23 g/mol
Boiling Pt: 195 °C (1013 hPa)
Melting Pt: –16 °C
Density: 0.8246 g/cm³ (20 °C)
Flash Pt: 81 °C
Storage Temperature: Ambient
MDL Number: MFCD00002988
CAS Number: 111-87-5
EINECS: 203-917-6
Merck Index: 13,06784
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7db4a6be3aedee4af43781ec6deb31631bc376c1&vid=80cb13f618a2d3181daf8310324993f1f7c69fb2&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR