(±)-γ-Octalactone ≥97% FCC, FG

Synonyms: (±)-Octan-4-olide, 5-Butyldihydrofuran-2(3H)-one

W279609-1KG-K W279609-100G-K W279609-5KG-K
220232-718EA 201.54 USD
220232-718 220232-717 220232-719
(±)-γ-Octalactone ≥97% FCC, FG
(±)-γ-Octalactone
Formula: C₈H₁₄O₂
MW: 142.2 g/mol
Boiling Pt: 234 °C (1013 hPa)
Density: 0.991 g/cm³ (0 °C)
MDL Number: MFCD00005402
CAS Number: 104-50-7
EINECS: 203-208-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR