Myristic acid Kosher

Warning

Synonyms: Tetradecylic acid, Tetradecanoic acid

W276413-10KG-K W276413-1KG-K W276413-25KG-K
220232-653EA 604.62 USD
220232-653 220232-654 220232-655
Myristic acid Kosher
Myristic acid
Formula: C₁₄H₂₈O₂
MW: 228.38 g/mol
Boiling Pt: 319 °C (1013 hPa)
Melting Pt: 51…54 °C
Density: 0.8622 g/cm³ (20 °C)
Flash Pt: >165 °C
Storage Temperature: Freezer
MDL Number: MFCD00002744
CAS Number: 544-63-8
EINECS: 208-875-2
Merck Index: 12,06416

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR